Key Documents
2636005
Supelco
Inlet Liner, Split/Splitless Type, Straight Design (wool packed)
Split (wool packed), pkg of 5 ea
About This Item
UNSPSC Code:
12352200
description
wool packed
packaging
pkg of 5 ea
L × O.D. × I.D.
72 mm × 6.3 mm × 3.4 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1075 and 1077 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)