Key Documents
2635805
Supelco
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
41115719
description
SPME/Splitless
packaging
pkg of 5 ea
L × O.D. × I.D.
74 mm × 6.35 mm × 0.75 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1075 and 1077 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)