Key Documents
2635801
Supelco
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
41115719
description
SPME/Splitless
packaging
pkg of 1 ea
manufacturer/tradename
Varian no equivalent (OEM)
L × O.D. × I.D.
74 mm × 6.35 mm × 0.75 mm
compatibility
configured for
for use with Varian 1075/1077 Injectors
technique used for SPME; Small volume
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1075 and 1077 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)