Key Documents
2633501
Supelco
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
41115719
packaging
pkg of 1 ea
L × O.D. × I.D.
99 mm × 5.0 mm × 0.75 mm
compatibility
for use with Shimadzu 14/15A/16 with SPL-14 Injectors
technique used for SPME
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Shimadzu (14, 15A, and 16) [with SPL-14 Injector]
Other Notes
I263350 --> material nos. 2633501 (1pk) & 2633505 (5pk).
Choose from one of the most recent versions:
Certificates of Analysis (COA)