Key Documents
2633405
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
41115719
packaging
pkg of 5 ea
manufacturer/tradename
Shimadzu 221-32544-00 (OEM)
L × O.D. × I.D.
99 mm × 5.0 mm × 3.0 mm
compatibility
for use with Shimadzu 14/15A/16 with SPL-14 Injectors
technique used for Trace analytes
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Shimadzu (14, 15A, and 16) [with SPL-14 Injector]
Other Notes
I263340 --> material nos. 2633405 (5pk) & 2633425 (25pk).
Choose from one of the most recent versions:
Certificates of Analysis (COA)