Key Documents
2633305
Supelco
Inlet Liner, Splitless Type, Single Taper Design (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
41115719
packaging
pkg of 5 ea
L × O.D. × I.D.
99 mm × 5.0 mm × 3.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Shimadzu (14, 15A, and 16) [with SPL-14 Injector]
Choose from one of the most recent versions:
Certificates of Analysis (COA)