Key Documents
2631405
Supelco
Inlet Liner, Direct Type, Straight Design (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
23151817
packaging
pkg of 5 ea
manufacturer/tradename
PerkinElmer N612-1006 (OEM, instrument manufacturer′s quartz liner)
L × O.D. × I.D.
86 mm × 4.0 mm × 1.0 mm
compatibility
for use with PerkinElmer Programmed Split/Splitless (PSS)
technique used for Trace analytes
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits PerkinElmer (PSS Injector)
Other Notes
I263140 --> material no. 2631405 (5pk)
Choose from one of the most recent versions:
Certificates of Analysis (COA)