Key Documents
2631305
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
12352200
packaging
pkg of 5 ea
L × O.D. × I.D.
86 mm × 4.0 mm × 2.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits PerkinElmer (PSS Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)