Key Documents
2631301
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
12352200
packaging
pkg of 1 ea
manufacturer/tradename
PerkinElmer N612-1004 (OEM, instrument manufacturer′s quartz liner)
L × O.D. × I.D.
86 mm × 4.0 mm × 2.0 mm
compatibility
for use with PerkinElmer Programmed Split/Splitless (PSS)
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits PerkinElmer (PSS Injector)
Other Notes
I263130 --> material nos. 2631301 (1pk), 2631305 (5pk), & 2631325 (25pk).
Footnote
Instrument manufacturer′s quartz liner.
Choose from one of the most recent versions:
Certificates of Analysis (COA)