2631205
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
technique used for SPME, pkg of 5 ea
About This Item
Recommended Products
packaging
pkg of 5 ea
L × O.D. × I.D.
92 mm × 6.35 mm × 0.75 mm
compatibility
configured for PerkinElmer Auto System, Model 9000 Split/Splitless Capillary Injector
technique used for SPME
General description
- Fits PerkinElmer (AutoSystem)
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)