Key Documents
2630301
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
41115719
packaging
pkg of 1 ea
L × O.D. × I.D.
100 mm × 5.0 mm × 4.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits PerkinElmer (2000 and 8000)
Choose from one of the most recent versions:
Certificates of Analysis (COA)