Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26250-U

SGE MicroVolume syringes with standard plunger, fixed needle

100F, volume 100 μL, needle size 25 ga, needle L 50 mm

Synonym(s):

SYRINGE SGE 100UL FN PT#2,25GA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

bevel tip stainless steel needle (point style 2)

needle size

25 ga

feature

needle type Fixed

packaging

pkg of 1 ea

manufacturer/tradename

SGE 005000

needle L

50 mm

volume

100 μL

Looking for similar products? Visit Product Comparison Guide

General description

Designers of syringes for microliter sample injections face two interesting problems: extremely fine plungers and proportionally large sample hold-up in the needle. SGE′s Plunger-in-Needle feature, which permits extremely small sample injections, and the SuperFlex plunger, which resists bending and kinking, addresses both problems. Other features of the SGE Microvolume Syringes [100F, volume 100 μL, needle size 25 ga (bevel tip), needle L 50 mm] also include:
  • Sharp and clear fused graduation.
  • Extended graduation lines and wide white backing for reduced errors.
  • Angled needle tip for optimum septum penetration.
  • Standard plunger is stainless steel individually fitted to its barrel.
  • Plunger Protection – An option on some SGE microvolume syringes.
  • Extended protection tube from the top of the flange helps prevent plunger bending during injection.
  • Rugged Guided Plunger – An option on some SGE microvolume syringes.
  • Extended barrel guides the plunger during injection and is suitable for training and student use.
  • Robust

SGE MicroVolume syringes with standard plunger, fixed needle are equipped with bevel tip needles, designed for optimum septum penetration and to prevent septum coring.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service