Key Documents
26249-U
SGE MicroVolume syringes with standard plunger, fixed needle
new
50F, volume 50 μL, needle size 25 ga, needle L 50 mm
Synonym(s):
SYRINGE SGE 50UL FN PT#2, 25GA
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
bevel tip stainless steel needle (point style 2)
needle size
25 ga
feature
needle type Fixed
packaging
pkg of 1 ea
manufacturer/tradename
SGE 004000
needle L
50 mm
volume
50 μL
General description
Designers of syringes for microliter sample injections face two interesting problems: extremely fine plungers and proportionally large sample hold-up in the needle. SGE′s Plunger-in-Needle feature, which permits extremely small sample injections, and the SuperFlex plunger, which resists bending and kinking, addresses both problems. Other features of the SGE Microvolume Syringes [50F, volume 50 μL, needle size 25 ga (bevel tip), needle L 50 mm] also include:
  • Sharp and clear fused graduation.
  • Extended graduation lines and wide white backing for reduced errors.
  • Angled needle tip for optimum septum penetration.
  • Standard plunger is stainless steel individually fitted to its barrel.
  • Plunger Protection – An option on some SGE microvolume syringes.
  • Extended protection tube from top of the flange helps prevent plunger bending during injection.
  • Rugged Guided Plunger – An option on some SGE microvolume syringes.
  • Extended barrel guides the plunger during injection and is suitable for training and student use.
  • Robust
SGE MicroVolume syringes with standard plunger, fixed needle are equipped with bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Choose from one of the most recent versions:
Certificates of Analysis (COA)