Key Documents
26235-U
SGE Plunger-Protected Syringe, Fixed Needle
volume 5 μL, needle size 26 ga, needle L 50 mm
Synonym(s):
SYRINGE SGE 5UL FN W/GUIDE, PT#2. 26GA
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
bevel tip stainless steel needle (point style 2)
needle size
26 ga
description
5F
feature
needle type Fixed
packaging
pkg of 1 ea
manufacturer/tradename
SGE 001000
needle L
50 mm
volume
5 μL
General description
Extended protection tube from top of the flange to help prevent plunger bending during injection.
Choose from one of the most recent versions:
Certificates of Analysis (COA)