Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness. This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).
Other Notes
This is a TightSpec column, which conforms to within +/- 6 mm of its stated length.
Kit Components Also Available Separately
Product No.
Description
SDS
21203-UEmpty Glass GC Column, for Agilent® 5880, 5890, 6890, 7820, and 7890 (Configuration "A"), L × O.D. × I.D. 3.3 ft (1.0 m) × 1/4 in. × 2.0 mm, TightSpecSDS
11880-UCoated GC Packing, phase 0.8% THEED, matrix 80/100 Carbopack C support, bottle of 15 gSDS
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.