Key Documents
25830
Swagelok® Connector to Male NPT
stainless steel, 1/2 in. Swagelok, 1/4 in. male NPT, 810-1-4
Synonym(s):
Swagelok® SS MALE CONNECTOR 1/2IN A TO 1/4IN B,SS-810-1-4
About This Item
UNSPSC Code:
41115717
eCl@ss:
32035302
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
810-1-4
fitting
1/2 in. Swagelok
1/4 in. male NPT
General description
Nuts and ferrules included.
Swagelok® connector to male NPT is a tube fitting, which delivers a leak-tight, gas-tight seal in an easy-to-install, disassemble and reassemble form. The robust tube grip with patented, two-ferrule technology is resistant to vibration fatigue and withstands high pressures and temperature extremes.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)