Key Documents
25807
Supelco
"T"-Type Gas Line Filter
stainless steel, 1/8 in. male
About This Item
UNSPSC Code:
41112500
material
stainless steel
fitting
1/8 in. male
General description
This gas line filter contains a replaceable 7 μm sintered stainless steel frit, and will remove dust and small particles that could otherwise foul downstream needle valves. Nuts and ferrules included.
Choose from one of the most recent versions:
Certificates of Analysis (COA)