Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25003

Supelco

HT-5 (aluminum clad) Capillary GC Column

L × I.D. 25 m × 0.32 mm, df 0.10 μm

Synonym(s):

HT-5, 25M .10UM .32MM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710

description

aluminum clad

parameter

10-460 °C temperature (isothermal)
10-480 °C temperature (programmed)

Beta value

800

df

0.10 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

25 m × 0.32 mm

matrix active group

Bonded; siloxane-carborane equivalent in polarity to poly(5% diphenyl/95% dimethyl siloxane) phase

application(s)

petroleum

column type

capillary non-polar

Looking for similar products? Visit Product Comparison Guide

General description

Application: This column offers the highest maximum temperature of any commercially available column. It is well suited for simulated distillation (Sim Dis) analyses of petroleum samples.
USP Code: None
Phase:
  • Bonded
  • Siloxane-carborane equivalent in polarity to poly(5% diphenyl/95% dimethyl siloxane)
Temp. Limits:
  • 10 °C to 460 °C (isothermal) or 480 °C (programmed)
Polysiloxane phase has high temperature stability, owing to its structural features, which are reactive terminal functional groups, degree of polymerization especially when the polysiloxane is phenyl-substituted, and the symmetrical substitution of different functional groups on the siloxane backbone.

Application

HT-5 (aluminum clad) Capillary GC Column may be used for the analysis of polar and high molecular weight compunds.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ha-Jung Kim et al.
Korean journal for food science of animal resources, 36(2), 206-214 (2016-05-20)
The aim of this study was to evaluate the potential use of fatty acids, triacylglycerols, and cholesterol in the detection of adulterated milk fat. The fatty acid, triacylglycerol, and cholesterol profiles of the mixtures of milk and non-milk fat (adulteration
High-temperature high-resolution gas chromatography: breaching the barrier to the analysis of polar and high molecular weight compounds.
Dos SP
TrAC, Trends in Analytical Chemistry, 18 (2), 126-136 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service