Norgren® Filter ideally removes oil vapor contaminants from compressed air. These contaminants can significantly shorten the lifetimes of downstream zero air generators, nitrogen generators, and other downstream instruments, and can affect the performance of the instruments. Oil Vapor-Removing Filter is recommended for use in any compressed line serving sensitive, high-performance gas chromatography or other laboratory instruments.
Legal Information
Norgren is a registered trademark of Norgren, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)