Norgren® particle filter removes liquids and solid particles as small as 5 microns in diameter and eliminates these contaminants from compressed air. The contaminants can significantly shorten the lifetimes of downstream zero air generators, nitrogen generators, and other downstream instruments, and can affect the performance of the instruments. One of these filters should be the initial filtering device downstream from the air compressor.
Legal Information
Norgren is a registered trademark of Norgren, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)