Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24990-U

Norgren® Filter

particle filter, pore size 40 μm, 1/4 in. female NPT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

material

particle filter

pore size

40 μm

fitting

1/4 in. female NPT

Looking for similar products? Visit Product Comparison Guide

General description

Norgren filter operating range is 40-1000 kPa.[1]

Legal Information

Norgren is a registered trademark of Norgren, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhalational anaesthesia in developing countries.
Ezi-Ashi TI
Anaesthesia, 38 (8), 736-747 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service