24917
Purge/Trap H
for use with Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting
Synonym(s):
PURGE TRAP H, Tekmar® 3000
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
application(s)
environmental
compatibility
for use with Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting
Looking for similar products? Visit Product Comparison Guide
General description
Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials, so that a broad range of polar and nonpolar, high and low molecular weight compounds can be trapped in a single tube. Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition. During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds. During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents.
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service