Key Documents
24817-U
Supelco
Merlin Microseal System (fits Varian®)
For 1079 injector; 1 nut, 1 inlet adapter, 1 o-ring, and 1 General Purpose (3-100 psi) septum
About This Item
UNSPSC Code:
12352200
NACRES:
SB.54
General description
The Merlin Microseal System is a septum-less system that provides very long life. Because there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner. The septum contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal. The septum can be used only with a syringe that has a 23 gauge "blunt tipped" needle, or with an SPME fiber assembly with a 23 gauge needle. The thicker shaft is required to make the necessary contact with the septum seals.
Application
Merlin Microseal System may be used to replace traditional septum to avoid leaking and bleeding caused during operating GC/MS system.[1] It may also be equipped to the injection port during GC analysis for determination of plasticizer N-butylbenzenesulfonamide and the pharmaceutical Ibuprofen in wastewater.[2]
Other Notes
Varian GCs require an inlet adapter and an o-ring in addition to the septum and nut. The septum incorporates a unique design with two sequential seals to provide a much longer life. Order replacement septa, or alternate versions, separately. Not compatible with the Varian 8200 autosampler.
Note: Do not use with beveled tips.
Legal Information
MicroSeal is a trademark of Merlin Instrument Co.
Varian is a registered trademark of Varian Medical Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Determination of the plasticizer N-butylbenzenesulfonamide and the pharmaceutical Ibuprofen in wastewater using solid phase microextraction (SPME).
Huppert N
Fresenius Journal of Analytical Chemistry, 326 (6), 529-536 (1998)
J M Diserens
Journal of AOAC International, 84(3), 853-860 (2001-06-22)
Although the use of pentachlorophenol (PCP) is prohibited in most countries, it is still widely found in the wood of pallets, containers, crates and in cardboard, paper, etc. Such material may contain not only PCP but other chlorophenols as well.