Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24700-U

Supelco

Needle valve for model 1067 air sampler, #3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

application(s)

industrial hygiene

compatibility

for use with 1067 Air Sampler

General description

Needle valve for model 1067 air sampler, #3 is used with Model 1067 dual channel ambient air sampler and provide stable flow rates for two independently controlled flow channels for sorbent tube sampling, over a range of 5-500mL/ min.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service