Key Documents
24667
Supelco
Critical Orifice For Model 1063 Bag Sampler
flow rate 1 mL/min
About This Item
UNSPSC Code:
41104008
packaging
pkg of 1 ea
parameter
1 mL/min flow rate
technique(s)
whole air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with 1063 and 1064 air sampler
Choose from one of the most recent versions:
Certificates of Analysis (COA)