24109
SP®-2380 Capillary GC Column
L × I.D. 15 m × 0.25 mm, df 0.20 μm
About This Item
Recommended Products
material
fused silica
agency
meets requirements for USP G48
parameter
≤25-275 °C temperature (isothermal or programmed)
Beta value
313
df
0.20 μm
technique(s)
gas chromatography (GC): suitable
L × I.D.
15 m × 0.25 mm
matrix active group
Stabilized; poly(90% biscyanopropyl/10% cyanopropylphenyl siloxane) phase
application(s)
agriculture
chemicals and industrial polymers
clinical
food and beverages
column type
capillary highly polar
Looking for similar products? Visit Product Comparison Guide
General description
USP Code: This column meets USP G48 requirements.
Phase:
- Stabilized
- Poly(90% biscyanopropyl/10% cyanopropylphenyl siloxane)
- Subambient to 275 °C (isothermal or programmed)
Application
- SP®-2380 Capillary Column may be suitable for determination of fatty acid composition in crude oil using capillary gas chromatography, used along with normal and reverse-phase high-performance liquid chromatography (HPLC) in order to study the content and composition of tocopherols, sterols, and phospholipids in soybean oils derived from genetically-modified soybeans.[1]
- It was also found suitable to be used for the identification of fatty acid methyl esters from mammalian samples by fast gas chromatography.[2]
- Furthermore, it is also suitable for determining the hydrogenation level and the level of isomerisation development during the hydrogenation process of edible oils by capillary gas chromatography.[3]
- Above all that it is also suitable to be used in gas chromatography equipped with flame ionization detector (FID) for analysing the cytotoxicity of fatty acids from seed oils containing conjugated linolenic acids (CLN) on mouse tumor and human monocytic leukemia cells.[4]
Other Notes
Legal Information
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service