23967
SGE Syringes for Other Autosamplers
needle size 26 ga, volume 10 μL, 10RX (removable needle), needle L × O.D. 50 mm × 0.47 mm, pkg of 6 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
bevel tip needle
glass syringe
needle size
26 ga
feature
point style 2
packaging
pkg of 6 ea
manufacturer/tradename
SGE 002180
needle L × O.D.
50 mm × 0.47 mm
volume
10 μL
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service