Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23946

SGE Microvolume Syringe for Rheodyne® and Valco® Valves, Fixed Needle

10FX-LC (Fixed needle), volume 10 μL, needle size 22 ga, needle L × O.D. 2 in. × 0.028 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

blunt tip stainless steel needle (point style 3)

needle size

22 ga

feature

needle type LTN (fixed hub)

packaging

pkg of 1 ea

manufacturer/tradename

SGE 002300

needle L × O.D.

2 in. × 0.028 in.

volume

10 μL

compatibility

for use with Rheodyne/Valco VISF-2 valve

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC
Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service