Key Documents
23883
In-Line Pressure Regulator
General Purpose version (outlet pressure 0-50 psi, 1/8 in. brass fittings)
About This Item
UNSPSC Code:
41112500
compatibility
Air, N2, Ar
General description
Install an in-line pressure regulator downstream of a gas cylinder regulator to adjust pressure closer to the point of use. 400 psig maximum input pressure. 1/8 in. Swagelok nuts and ferrules included.
Not recommend to be used with a carrier or fuel gas for a GC because of the neoprene diaphragm.
Application
Air, H2, N2, Ar
Choose from one of the most recent versions:
Certificates of Analysis (COA)