Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23858

Supelco

Glass Packed GC Column (fits Varian®)

phase 3% SP-2100, matrix 100/120 SUPELCOPORT support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

glass column

manufacturer/tradename

Varian

parameter

0-350 °C temperature (isothermal or programmed)

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

6.0 ft (1.8 m) × 1/4 in. × 2.0 mm

matrix

100/120 SUPELCOPORT support

matrix active group

3% SP-2100 phase

column type

packed GC

compatibility

for use with Varian

Looking for similar products? Visit Product Comparison Guide

General description

GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.[1]
Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness. This column fits Varian 3000, 4000, 6000, and Vista (FID, inj A to det A).

Application

Glass GC column packed with 3% SP-2100 on Supelcoport 100/120 was used for gas chromatographic analysis of synthetic esters of pregnenolone, during characterization of the lipoidal derivatives of pregnenolone which was prepared by incubation of the steroid with adrenal mitochondria.[2]

Legal Information

Varian is a registered trademark of Varian Medical Systems, Inc.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mellon-Nussbaum et al.
The Journal of biological chemistry, 254(24), 12500-12505 (1979-12-25)
Using mass spectrometric, radioisotopic, chromatographic and chemical techniques, five fatty acid esters of 3 beta-hydroxy-5-pregnen-20-one (pregnenolone) have been identified as components of the lipoidal derivatives biosynthesized in vitro with bovine adrenal mitochondria. The five compounds are: pregnenolone arachidonate, pregnenolone linoleate
Scott RPW.
Introduction to Analytical Gas Chromatography (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service