Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23832-U

In-Line Pressure Regulator (economy model)

Regulator body only (outlet pressure 0-50 psi)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

parameter

300 psig max. inlet pressure (21 bar)

Looking for similar products? Visit Product Comparison Guide

General description

The economy in-line pressure regulator is an inexpensive version for general lab applications. Use it with gas streams for pneumatic control, and other non-chromaotgraphic purposes. 300 psig maximum inlet pressure. 1/8 in. brass Swagelok nuts and ferrules included.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service