Key Documents
23815
Supelco
Capillary Cleaving Tool
Replacement Blade (for P/N 23814)
About This Item
UNSPSC Code:
41115700
material
sapphire cutting blade
packaging
pkg of 1 ea
General description
Capillary Cleaving Tool increases the column performance by making the properly sized, squarely-cut ferrule to seal the capillary column end.
This handy tool makes scalpel-like cuts to polyimide-coated fused silica, leaving no jagged edges to create problems. The industrial sapphire cutting edges remains sharp indefinitely.
Other Notes
Replacement sapphire cutting blade for P/N 23814.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)