Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23811-U

Supelco

Drill Bits

diam. 0.35 mm, pkg of 6 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813
NACRES:
SB.54

packaging

pkg of 6 ea

diam.

0.35 mm

compatibility

for use with Pin Vise Drill Kit

Looking for similar products? Visit Product Comparison Guide

Application

Drill bits may be used to drill a hole through a solid pellet, during thermionic emission flame gas chromatography, used for the determination of residues of di-syston and its metabolites in various crops and products.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of residues of Di-Syston and metabolites by thermionic flame gas chromatography.
Thornton JS and Anderson CA.
Journal of Agricultural and Food Chemistry, 16 (6), 895-898 (1968)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service