Key Documents
23639
Supelco
Capillary Starter Kit
About This Item
UNSPSC Code:
41115700
packaging
pkg of 1 kit
storage temp.
−20°C
General description
This convenient kit includes all the tools needed for installing capillary GC columns and related accessories. It contains:
6 in. (15 cm) pipe cleaners
Tweezers
Pocket mirror with rotating head
1/4in. × 5/16in. open end wrench
Capillary Cleaving Tool
6 in. (15 cm) stainless steel ruler
Small flashlight
Pin vise drill kit
Screw-type septum puller
Choose from one of the most recent versions:
Certificates of Analysis (COA)