Key Documents
23633
Supelco
Reducing Union (1/4 in. to 1/16 in.)
1 stainless steel reducing union 1/4 in. to 1/16 in.
About This Item
UNSPSC Code:
41115700
General description
Reducing unions are mostly suitable as metal column terminators. All original reducing unions have a bore in the centre of about 6mm in length and an internal diameter of 1.2mm. Moreover, reducing unions have a large bore at the non-reducing site. Modification is necessary in reducing union in order to minimize peak broadening, pressure resistance and to ease the installation process.[1]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Instrumentation for High Performance Liquid Chromatography.
Huber JFK
Science, 84-88 (2011)