Key Documents
23626
Supelco
Solvent Rinse Kit
About This Item
UNSPSC Code:
23151816
packaging
pkg of 1 kit
General description
Solvent rinse kits are generally suitable to rinse a column. The detector end of the column is placed in the rinse kit vial through the straight arm. Solvent is added to the vial supplying inert gas from the other arm of tee-fitting. Pressure created in the sealed vial forces the solvent to pass through the column rinsing the column.[1]
Application
Solvent rinse kit was used in column coating procedure while investigating oligoproline chiral selectors for enantioselective gas chromatography.[2] It may be used in rinsing dried packing capillary entrapped column used for HPLC and capillary electrochromatography.[3]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Chirica et al.
Analytical chemistry, 72(15), 3605-3610 (2000-08-22)
A new design of immobilized particle separation media for capillary liquid chromatography and electrochromatography has been developed. A mixture of porogenic solvents and methacrylate-based monomers is pumped through a packed column to provide, following a polymerization step, an organic matrix
Dean Rood
The Troubleshooting and Maintenance Guide for Gas Chromatographers, 222-223 (2007)
Min Li et al.
Journal of chromatography. A, 1191(1-2), 199-204 (2008-02-12)
A new chiral stationary phase for gas chromatography was prepared by covalently attaching a diproline chiral selector that has proven to be effective in liquid chromatography to a methylhydrosiloxane-dimethylsiloxane copolymer. With this new chiral stationary phase for GC, racemic aromatic