Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23613

Chart Paper for Recorder

PerkinElmer C005-0610 (OEM), chart type, 0-100 English, pkg of 1 roll

Synonym(s):

CHART PAPER PE R100, 0-100,1 ROLL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
14111500

feature

chart type 0-100 English

packaging

pkg of 1 roll

manufacturer/tradename

PerkinElmer C005-0610 (OEM)

compatibility

for use with PerkinElmer R100

Looking for similar products? Visit Product Comparison Guide

General description

Main purpose of a recorder is to record electrical and non-electrical quantity as a function of time. Graphic recorders use elementary rudiments like chart paper for displaying and storing the physical quantity and pen for marking the variation in physical quantity. Chart paper is passed through the recorder at constant rate. The recorder itself becomes a device for measuring retention time. A certain number of inches or centimetres of chart paper corresponds to a certain number of minutes.

Application

Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service