Key Documents
23609
Chart Paper for Recorder
Kipp & Zonen XR-18 (OEM), chart paper L × W 66 ft × 230 mm, chart type, 100-0 English, pkg of 1 roll
About This Item
UNSPSC Code:
14111500
feature
chart type 100-0 English
packaging
pkg of 1 roll
manufacturer/tradename
Kipp & Zonen XR-18 (OEM)
WESTRONICS
chart paper L × W
66 ft × 230 mm
compatibility
for use with Kipp & Zonen BD 111/112
General description
Main purpose of a recorder is to record electrical and non-electrical quantity as a function of time. Graphic recorders use elementary rudiments like chart paper for displaying and storing the physical quantity and pen for marking the variation in physical quantity.[1] Chart paper is passed through the recorder at constant rate. The recorder itself becomes a device for measuring retention time. A certain number of inches or centimetres of chart paper corresponds to a certain number of minutes.[2]
Application
Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.[3]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)