23493
CapSeal Bullet Capillary Ferrule, Long Design
I.D. 0.4 mm, 0.10-0.25 mm Column I.D., pkg of 48 ea
About This Item
Recommended Products
feature
ferrule type long
packaging
pkg of 48 ea
I.D.
0.4 mm
compatibility
0.10-0.25 mm Column I.D.
Looking for similar products? Visit Product Comparison Guide
General description
Characteristics: Seals at 1/8 turn past finger tight. Reusable. A special end taper reduces graphite extrusion into fitting.
Material Composition: graphite in an aluminum base
Maximum Temperature: 450 °C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service