Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23445

Supelco

Branched paraffins

15 ppm each component in nitrogen, cylinder of 14 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12181503

grade

analytical standard

form

gas

packaging

cylinder of 14 L

manufacturer/tradename

Scott Gas T2-14

availability

not available in EU, Canada

concentration

15 ppm each component in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

  • Branched paraffins, 15 ppm each component in nitrogen, is a multi-component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.

Analyte

Description

    2,2-Dimethylbutane
    2,2-Dimethylpropane
    2-Methylbutane
    2-Methylpentane
    3-Methylpentane
    2-Methylpropane

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service