23438
Carbon Monoxide, Carbon Dioxide, Hydrogen and Oxygen
0.5 % (w/w) each component in nitrogen, cylinder of 14 L, analytical standard
About This Item
Recommended Products
grade
analytical standard
form
gas
packaging
cylinder of 14 L
manufacturer/tradename
Scott Gas T217-14
availability
not available in Canada, EU
concentration
0.5 % (w/w) each component in nitrogen
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
multi-component solution
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
- Carbon Monoxide, Carbon Dioxide, Hydrogen and Oxygen, 0.5% (w/w) each component in nitrogen, is a multi-component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers.
- A certificate of analysis is included.
- Packaged in lightweight, non-refillable, recyclable containers.
- Portable, convenient, and easy to use.
- Manufactured gravimetrically for maximum accuracy.
- NIST traceable.
Analyte
regulator
signalword
Danger
hcodes
Hazard Classifications
Repr. 1A
Storage Class
2A - Gases
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service