Key Documents
23413-U
Supelco
Gold-Plated Inlet Seal (Cross Design)
pkg of 2 ea
About This Item
UNSPSC Code:
41121815
NACRES:
SB.54
material
stainless steel (gold plated cross design)
packaging
pkg of 2 ea
General description
The inlet seal on an Agilent GC must be regularly changed to prevent sample adsorption due to accumulation of sample residue and/or septum fragments. Supelco manufactures replacement inlet seals using a high quality stainless steel plated with pure gold to insure inertness. No brighteners are used in the plating process. This cross design seal is suitable for applications with high split flows (>200 mL/min.). Each pack includes one washer for each seal.
Application
Gold-plated inlet seal may be used in the GC injection port for thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS), where residual accumulation was noticed resulting in analyte adsorption and elevating signal background. Hence, replacing the gold-plated seal in regular intervals decreased the interference.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Precautions for in-injection port thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS) as applied to aerosol filter samples.
Ho, Steven Sai Hang, et al.
Atmospheric Environment, 45, 1491-1496 (2011)