Key Documents
23312
Supelco
Capillary Column Nut Kit (fits Shimadzu)
About This Item
UNSPSC Code:
41121815
packaging
pkg of 1 ea
General description
Kit includes stainless steel nut, spring, stainless steel front ferrule, and metal back spacer. Note that 2 kits are needed per column.
Legal Information
Shimadzu is a trademark of Shimadzu Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)