Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23093

Supelco

Glass Packed GC Column (fits Agilent, Configuration "A")

phase SP®-1000, 1%, matrix 60/80 Carbopack B support, L × O.D. × I.D. 6.6 ft (2.0 m) × 1/4 in. × 2.0 mm, TightSpec

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

glass column

manufacturer/tradename

Agilent

parameter

0-225 °C temperature (isothermal or programmed)

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

6.6 ft (2.0 m) × 1/4 in. × 2.0 mm, TightSpec

matrix

60/80 Carbopack B support

matrix active group

SP®-1000 phase, 1%

column type

packed GC

compatibility

for use with Agilent Configuration A

Looking for similar products? Visit Product Comparison Guide

General description

In a packed column, the carrier gas percolates through the porous bed and separation is achieved by moving the solutes apart. Packed columns are made up of two main materials; stainless steel and Pyrex glass. Pyrex glass columns are shaped as desired by coiling at about 700°C. Glass column are in some cases treated with silanizing reagent to eliminate the surface hydroxyl groups which becomes catalytically active and produce asymmetric peaks. Carbopack B, better known as graphitized carbon black has nonporous, nonpolar and homogeneous surface. This adsorbent is suitable as stationary phase in both packed and capillary columns. Carbopack B cartridges are more efficient than C-18 for extracting phenols.
Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness. This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).

Application

Carbopack B packed glass column was used in GC analysis of C2-C5 fatty acids in aqueous media.

Other Notes

This is a TightSpec column, which conforms to within +/- 6 mm of its stated length.

Legal Information

SP is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatographic analysis of C2-C5 fatty acids in aqueous media using Carbopack B-Carbowax 20M-phosphoric acid.
Du Preez, J. C., and P. M. Lategan
Journal of Chromatography A, 150, 259-262 (1978)
Raymond P.W. Scott
Introduction to Analytical Gas Chromatography (1997)
A Di Corcia et al.
Analytical chemistry, 63(6), 580-585 (1991-03-15)
A general liquid-solid extraction procedure for the isolation of pesticides from groundwater and drinking water for high-performance liquid chromatography (HPLC) is presented. This simple and rapid procedure involved passing a 2-L sample through a 250-mg graphitized carbon black (Carbopack B)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service