Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23077

Supelco

Glass Packed GC Column (fits Agilent, Configuration "A")

phase 1.5% SP-2250 + 1.95% SP-2401, matrix 100/120 SUPELCOPORT support, L × O.D. × I.D. 6.6 ft (2.0 m) × 1/4 in. × 4.0 mm, TightSpec

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

glass column

manufacturer/tradename

Agilent

parameter

0-250 °C temperature (isothermal or programmed)

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

6.6 ft (2.0 m) × 1/4 in. × 4.0 mm, TightSpec

matrix

100/120 SUPELCOPORT support

matrix active group

1.5% SP-2250 + 1.95% SP-2401 phase

column type

packed GC

compatibility

for use with Agilent Configuration A

Looking for similar products? Visit Product Comparison Guide

General description

In a packed column, the carrier gas percolates through the porous bed and separation is achieved by moving the solutes apart. Packed columns are made up of two main materials; stainless steel and Pyrex glass. Pyrex glass columns are shaped as desired by coiling at about 700°C. Glass column are in some cases treated with silanizing reagent to eliminate the surface hydroxyl groups which becomes catalytically active and produce asymmetric peaks.[1]
Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness. This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).

Other Notes

This is a TightSpec column, which conforms to within +/- 6 mm of its stated length.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond P.W. Scott
Introduction to Analytical Gas Chromatography (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service