Key Documents
23020-U
Supelco
HPLC sample loop 100μl ss 7000psi ea
sample loop volume 100 μL
About This Item
UNSPSC Code:
41115854
NACRES:
SB.51
material
stainless steel sample loop and fittings
manufacturer/tradename
VICI Valco® SL100CUW
sample loop volume
100 μL
fitting
10-32 Valco fittings (nuts and ferrules included)
compatibility
for use with Valco HPLC valve
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)