Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23019-U

Supelco

HPLC sample loop 50μl ss 7000psi ea

sample loop volume 50 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

material

stainless steel sample loop and fittings

manufacturer/tradename

VICI Valco® SL50CUW

sample loop volume

50 μL

fitting

10-32 Valco fittings (nuts and ferrules included)

compatibility

for use with Valco HPLC valve

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service