22950
6-Port Sampling and Switching Valve
1/16 in. fitting, maximum temperature 300 °C
All Photos(2)
About This Item
UNSPSC Code:
41112500
eCl@ss:
32060101
Recommended Products
feature
number of ports 6
manufacturer/tradename
VICI Valco®
maximum temperature
300 °C
fitting
1/16 in. fitting
General description
Sampling valve basically supports the discrete volumes of the treated samples stream to periodically inject into the carrier gas stream of the chromatograph. Sampling valves are fitted with electric or pneumatic actuators which help them to be operated automatically. Switching valve are critical if binary pumps are used. It allows employment of different desired mobile phases. Switching valve consists of five parts: control unit, connecting device, valve body, a rotor and preloaded assembly.
Application
Foreflushing allows you to separate low molecular weight compounds. In the example shown, C1-C5 hydrocarbons are rapidly eluted from Column 1 onto Column 2 (position A). When the valve is switched to position B, C6 and heavier compounds (which have moved slowly through Column 1) are eluted from Column 1 to the detector. C1-C5 hydrocarbons are eluted from Column 2 back onto Column 1, then to the detector.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Satinder Ahuja
Chiral Separation Methods for Pharmaceutical and Biotechnological Products, 218-218 (2011)
Walt Boyes
Instrumentation Reference Book, 414-414 (2009)