Key Documents
22916
8-Port Sampling and Switching Valve
1/8 in. fitting, maximum temperature 300 °C
About This Item
UNSPSC Code:
41112500
feature
number of ports 8
manufacturer/tradename
VICI Valco®
maximum temperature
300 °C
fitting
1/8 in. fitting
General description
Sampling valve basically supports the discrete volumes of the treated samples stream to periodically inject into the carrier gas stream of the chromatograph. Sampling valves are fitted with electric or pneumatic actuators which help them to be operated automatically.[1] Switching valve are critical if binary pumps are used. It allows employment of different desired mobile phases. Switching valve consists of five parts: control unit, connecting device, valve body, a rotor and preloaded assembly.[2]
Select between two columns connected to a single detector requiring auxiliary gas for the second column. The figure shows the 8-port sampling valve with two sample loops used for repetitive sample injections on one column.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Walt Boyes
Instrumentation Reference Book, 414-414 (2009)
Satinder Ahuja
Chiral Separation Methods for Pharmaceutical and Biotechnological Products, 218-218 (2011)