Key Documents
22887
Chart Paper for Recorder
chart type, blank, Z-fold black
About This Item
UNSPSC Code:
14111500
material
Z-fold black
feature
chart type blank
manufacturer/tradename
Agilent/HP 9270-0658 (OEM)
Waters 74703 (OEM)
compatibility
for use with Agilent/HP 1080, 3380, 3388A, 5380, 5880
for use with PerkinElmer Sigma Series (except Sigma 15
for use with Waters recorders
General description
Main purpose of a recorder is to record electrical and non-electrical quantity as a function of time. Graphic recorders use elementary rudiments like chart paper for displaying and storing the physical quantity and pen for marking the variation in physical quantity.[1] Chart paper is passed through the recorder at constant rate. The recorder itself becomes a device for measuring retention time. A certain number of inches or centimetres of chart paper corresponds to a certain number of minutes.[2]
Application
Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.[3]
Other Notes
Equivalent to Manufacturer′s Chart No. 9270-0658.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)